Assay Details
| Gene Symbol: Ddo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027442.5 | NP_081718.2 | 1 - 2 | 79 | 61 | |
| GenBank mRNA | AK017475.1 | - | 1 - 2 | 45 | 61 | |
| AK019898.1 | - | 1 - 2 | 68 | 61 | ||
| AK033375.1 | - | 1 - 2 | 79 | 61 | ||
| AK085947.1 | - | 1 - 2 | 56 | 61 | ||
| AK161548.1 | - | 1 - 2 | 37 | 61 | ||
| AK165344.1 | - | 48 | 61 | |||
| BC006690.1 | - | 1 - 2 | 31 | 3582980 | 61 | |
| BC027312.1 | - | 1 - 2 | 33 | 4007445 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK017475.1, AK019898.1, AK033375.1, AK085947.1, AK161548.1, AK165344.1, BC006690.1, BC027312.1, NM_027442.5 transcripts.
Set Membership: |
|

Ddo
Genomic Map