Assay Details
| Gene Symbol: Trmt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014181.1 | - | 3 - 4 | 223 | 165 | |
| BC048703.1 | - | 3 - 4 | 207 | 6509593 | 165 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC048703.1 transcripts.
Set Membership: |
|

Trmt11
Genomic Map