Assay Details
| Gene Symbol: Trmt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014181.1 | - | 4 - 5 | 325 | 153 | |
| BC048703.1 | - | 4 - 5 | 309 | 6509593 | 153 | |
| BY259542.1 | - | 4 - 5 | 154 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC048703.1 transcripts.
Set Membership: |
|

Trmt11
Genomic Map