Assay Details
| Gene Symbol: Mtmr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028860.2 | NP_083136.2 | 14 - 15 | 1971 | 90 | |
| GenBank mRNA | AK141138.1 | - | 14 - 15 | 1951 | 90 | |
| BC032166.1 | - | 14 - 15 | 1954 | 5352919 | 90 | |
| BC081544.1 | - | 14 - 15 | 1955 | 3672099 | 90 | |
| CF532066.1 | - | 2 - 3 | 313 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mtmr3
Genomic Map