Assay Details
| Gene Symbol: Mtmr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028860.2 | NP_083136.2 | 16 - 17 | 3514 | 87 | |
| GenBank mRNA | AK007036.1 | - | 1 - 2 | 263 | 87 | |
| BC026586.1 | - | 1 - 2 | 1306 | 87 | ||
| BC081544.1 | - | 16 - 17 | 3498 | 3672099 | 87 | |
| CX566279.1 | - | 1 - 2 | 448 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mtmr3
Genomic Map