Assay Details
| Gene Symbol: Atp6v1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134157.2 | NP_598918.1 | 6 - 7 | 657 | 110 | |
| GenBank mRNA | AF435091.1 | - | 6 - 7 | 657 | 110 | |
| AK052604.1 | - | 6 - 7 | 628 | 110 | ||
| AK052707.1 | - | 6 - 7 | 619 | 110 | ||
| AK078810.1 | - | 6 - 7 | 685 | 110 | ||
| AK085549.1 | - | 6 - 7 | 658 | 110 | ||
| AK168980.1 | - | 6 - 7 | 685 | 110 | ||
| BC004789.1 | - | 3 - 4 | 251 | 110 | ||
| BC017127.1 | - | 6 - 7 | 642 | 4489144 | 110 | |
| BC062202.1 | - | 6 - 7 | 616 | 2650839 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp6v1b1
Genomic Map