Assay Details
| Gene Symbol: Slc25a26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017037.1 | - | 3 - 4 | 212 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a26| Gene Symbol: Slc25a26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017037.1 | - | 3 - 4 | 212 | 96 | |
Set Membership: |
|