Assay Details
| Gene Symbol: Aplf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170489.1 | NP_001163960.1 | 1 - 2 | 312 | 134 | |
| GenBank mRNA | AK008513.1 | - | 1 - 2 | 313 | 134 | |
| AK078805.1 | - | 1 - 2 | 274 | 134 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK008513.1, AK078805.1, NM_001170489.1 transcripts.
Set Membership: |
|

Aplf
Genomic Map