Assay Details
Gene Symbol: Vmn1r74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134206.1 | NP_598967.1 | 1 | 868 | 128 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P332506 | NM_134206.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P332506 | NM_134206.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122051 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P332506 | NM_134206.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P332506 | NM_134206.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332506 | NM_134206.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P332506 | NM_134206.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969798 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969790 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969794 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969802 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969799 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969780 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969788 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969787 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969789 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969805 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969792 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969785 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969782 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969784 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P122051 | - | - |
Whole Mouse Genome G4122A | A_51_P332506 | NM_134206.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P332506 | NM_134206.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114965 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137093 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P122051 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06617861 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06002024 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06002026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969812 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969845 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969818 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969811 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969816 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969835 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969828 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969820 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969840 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969794 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969799 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969848 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969801 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969813 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969809 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969824 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969768 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969776 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969806 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969826 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969805 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969792 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969830 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969838 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969842 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969815 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969813 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969831 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969792 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969799 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969830 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969826 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969780 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06002022 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969790 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969798 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969787 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969812 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969835 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969776 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06247872 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969788 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969829 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969824 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969828 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969789 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969809 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06165286 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969806 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06643319 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969833 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06058709 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969820 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969808 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969785 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969840 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969815 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06501522 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969837 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969784 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969816 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969805 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969807 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969794 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969838 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969811 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969818 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24969782 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10549985 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10549984 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 538961 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2060692 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2060692 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 101570438 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002060692 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2463192 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2463192 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1230014 | - | - |
mouse-6 v1.1 (Illumina) | scl9723.1.1_216-S | - | - |
mouse-6 v1.1 (Illumina) | GI_21717734-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 101570438 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2060692 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl9723.1.1_216-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2060692 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101570438 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2838831 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2838831 | - | - |
