Assay Details
| Gene Symbol: Vmn1r73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134203.1 | NP_598964.1 | 1 | 744 | 136 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1977169 | NM_134203.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138135 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1977169 | NM_134203.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P130233 | NM_134203.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969503 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969504 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969486 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969510 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969522 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969483 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969488 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24969490 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138135 | - | - |
| Whole Mouse Genome G4122A | A_51_P130233 | NM_134203.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123793 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138135 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06572893 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06330620 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06039657 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06039658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24969533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969540 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06527354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05944749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969538 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969480 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969489 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969488 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06135723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06039661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969525 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06458130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06039651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969486 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969537 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969520 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24969494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06031776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05995457 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10549983 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10549982 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 632496 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4670497 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4670497 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004670497 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2467955 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2467955 | - | - |
| mouse-6 v1.1 (Illumina) | scl9724.1.1_3-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4670497 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl9724.1.1_3-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4670497 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2467955 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2467955 | - | - |

Vmn1r73
Genomic Map