Assay Details
| Gene Symbol: Emid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080595.2 | NP_542162.1 | 10 - 11 | 973 | 147 | |
| GenBank mRNA | AJ416093.1 | - | 10 - 11 | 973 | 147 | |
| AK153702.1 | - | 11 - 12 | 1092 | 147 | ||
| BC130023.1 | - | 10 - 11 | 978 | 40140754 | 147 | |
More Information
Updated Mapping Information
Set Membership: |
|

Emid1
Genomic Map