Assay Details
| Gene Symbol: Emid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC012248.1 | - | 4 - 5 | 225 | 60 | |
| BC021849.1 | - | 3 - 4 | 127 | 60 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Emid1| Gene Symbol: Emid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC012248.1 | - | 4 - 5 | 225 | 60 | |
| BC021849.1 | - | 3 - 4 | 127 | 60 | ||
Set Membership: |
|