Assay Details
| Gene Symbol: Echdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110195.1 | NP_001103665.1 | 1 - 2 | 130 | 107 | |
| GenBank mRNA | AK150932.1 | - | 1 - 2 | 114 | 107 | |
| AK152285.1 | - | 1 - 2 | 114 | 107 | ||
| AK153454.1 | - | 1 - 2 | 114 | 107 | ||
| BY325699.1 | - | 1 - 2 | 131 | 107 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK150932.1, AK152285.1, AK153454.1, NM_001110195.1 transcripts.
Set Membership: |
|

Echdc1
Genomic Map