Assay Details
| Gene Symbol: Slc6a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172890.3 | NP_766478.1 | 10 - 11 | 1390 | 90 | |
| GenBank mRNA | AK053078.1 | - | 10 - 11 | 1391 | 90 | |
| AK053261.1 | - | 10 - 11 | 1232 | 90 | ||
| AK140423.1 | - | 10 - 11 | 1393 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a11
Genomic Map