Assay Details
| Gene Symbol: Nop2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138747.2 | NP_620086.2 | 1 - 2 | 117 | 82 | |
| GenBank mRNA | AK079914.1 | - | 1 - 2 | 91 | 82 | |
| AK154006.1 | - | 1 - 2 | 71 | 82 | ||
| AK156568.1 | - | 1 - 2 | 64 | 82 | ||
| BC007151.1 | - | 1 - 2 | 61 | 3592148 | 82 | |
| BY244994.1 | - | 1 - 2 | 117 | 82 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK154006.1,BC007151.1,NM_138747.2 transcripts.
This assay is in the 5'UTR region of the AK154006.1,BC007151.1,NM_138747.2 transcripts.
Set Membership: |
|

Nop2
Genomic Map