Assay Details
| Gene Symbol: Fam184a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081428.1 | NP_001074897.1 | 10 - 11 | 2344 | 67 | |
| GenBank mRNA | AK139673.1 | - | 5 - 6 | 835 | 67 | |
More Information
Set Membership: |
|

Fam184a
Genomic Map