Assay Details
| Gene Symbol: Rfx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159389.1 | NP_001152861.1 | 9 - 10 | 1011 | 113 | |
| NM_177306.3 | NP_796280.1 | 6 - 7 | 642 | 113 | ||
| GenBank mRNA | AK029809.1 | - | 6 - 7 | 643 | 113 | |
| AK049380.1 | - | 9 - 10 | 991 | 113 | ||
| BC132104.1 | - | 4 - 5 | 325 | 40130381 | 113 | |
| BC132294.1 | - | 4 - 5 | 325 | 40130571 | 113 | |
| BC145065.1 | - | 4 - 5 | 325 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rfx6
Genomic Map