Assay Details
| Gene Symbol: Rfx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159389.1 | NP_001152861.1 | 10 - 11 | 1062 | 104 | |
| NM_177306.3 | NP_796280.1 | 7 - 8 | 693 | 104 | ||
| GenBank mRNA | AK029809.1 | - | 7 - 8 | 694 | 104 | |
| AK049380.1 | - | 10 - 11 | 1042 | 104 | ||
| BC132104.1 | - | 5 - 6 | 376 | 40130381 | 104 | |
| BC132294.1 | - | 5 - 6 | 376 | 40130571 | 104 | |
| BC145065.1 | - | 5 - 6 | 376 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rfx6
Genomic Map