Assay Details
| Gene Symbol: Rfx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159389.1 | NP_001152861.1 | 12 - 13 | 1369 | 101 | |
| NM_177306.3 | NP_796280.1 | 9 - 10 | 1000 | 101 | ||
| GenBank mRNA | AK029809.1 | - | 9 - 10 | 1001 | 101 | |
| AK049380.1 | - | 12 - 13 | 1349 | 101 | ||
| BC132104.1 | - | 7 - 8 | 683 | 40130381 | 101 | |
| BC132294.1 | - | 7 - 8 | 683 | 40130571 | 101 | |
| BC145065.1 | - | 7 - 8 | 683 | 101 | ||
| FJ804414.1 | - | 11 - 12 | 1217 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rfx6
Genomic Map