Assay Details
| Gene Symbol: Catsper4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130030.1 | NP_001123502.1 | 5 - 6 | 566 | 64 | |
| NM_177866.4 | NP_808534.1 | 7 - 8 | 785 | 64 | ||
| GenBank mRNA | AK077145.1 | - | 7 - 8 | 772 | 64 | |
| BC108974.2 | - | 7 - 8 | 765 | 40047424 | 64 | |
| BC108975.1 | - | 7 - 8 | 765 | 40047425 | 64 | |
| EU781510.1 | - | 6 - 7 | 662 | 64 | ||
| EU781511.1 | - | 5 - 6 | 541 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Catsper4
Genomic Map