Assay Details
| Gene Symbol: Catsper4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130030.1 | NP_001123502.1 | 7 - 8 | 946 | 91 | |
| NM_177866.4 | NP_808534.1 | 9 - 10 | 1165 | 91 | ||
| GenBank mRNA | AK077145.1 | - | 9 - 10 | 1152 | 91 | |
| BC108974.2 | - | 9 - 10 | 1145 | 40047424 | 91 | |
| BC108975.1 | - | 9 - 10 | 1145 | 40047425 | 91 | |
| EU781510.1 | - | 8 - 9 | 1042 | 91 | ||
| EU781511.1 | - | 7 - 8 | 921 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Catsper4
Genomic Map