Assay Details
| Gene Symbol: Man1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008548.4 | NP_032574.1 | 4 - 5 | 1333 | 131 | |
| GenBank mRNA | AK028996.1 | - | 4 - 5 | 1248 | 131 | |
| AK031038.1 | - | 4 - 5 | 1334 | 131 | ||
| AK042371.1 | - | 4 - 5 | 1269 | 131 | ||
| AK043932.1 | - | 4 - 5 | 1274 | 131 | ||
| AK148407.1 | - | 4 - 5 | 1274 | 131 | ||
| AK154820.1 | - | 4 - 5 | 1275 | 131 | ||
| AK172376.1 | - | 4 - 5 | 1276 | 131 | ||
| BC015265.1 | - | 4 - 5 | 1234 | 4223319 | 131 | |
| U04299.1 | - | 3 - 4 | 937 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Man1a
Genomic Map