Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000001081.mm.1 |
NM_001198635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001081.mm.1 |
NM_001198635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001081.mm.1 |
NM_008635.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001081.mm.1 |
NM_008635.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001081.mm.1 |
TC1000000150.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000001079.mm.1 |
NM_001198635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001079.mm.1 |
NM_001198635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001079.mm.1 |
NM_008635.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001079.mm.1 |
NM_008635.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001079.mm.1 |
TC1000000150.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000001080.mm.1 |
NM_001198635.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001080.mm.1 |
NM_001198635.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001080.mm.1 |
NM_008635.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001080.mm.1 |
NM_008635.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000001080.mm.1 |
TC1000000150.mm.1 |
MTA TC |