Assay Details
| Gene Symbol: 4930598N05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016412.1 | - | 1 - 2 | 517 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | syntaxin binding protein 5 (tomosyn) | Stxbp5 |
0710001E20Rik, 4930565N16Rik, AW742610, LGL3, mKIAA1006 |
-333.8kb | Anti-sense | Chr. 10 - 9755547 - 9901227 on NCBI Build GRCm38 |
| Downstream | androglobin | Adgb | 9130014G24Rik | 81.5kb | Anti-sense | Chr. 10 - 10335703 - 10475135 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

4930598N05Rik
Genomic Map