Assay Details
| Gene Symbol: 4930598N05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016412.1 | - | 3 - 4 | 742 | 135 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | syntaxin binding protein 5 (tomosyn) | Stxbp5 |
0710001E20Rik, 4930565N16Rik, AW742610, LGL3, mKIAA1006 |
-356.3kb | Anti-sense | Chr. 10 - 9755547 - 9901227 on NCBI Build GRCm38 |
| Downstream | androglobin | Adgb | 9130014G24Rik | 77.0kb | Anti-sense | Chr. 10 - 10335703 - 10475135 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2360845 | AK132706.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 674867 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 922966 | AB038011.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 922966 | AB041597.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 922966 | AJ279796.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 922966 | AJ279797.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 922966 | AK088052.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 922966 | NM_020285.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 922966 | NM_138631.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 120386 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 870352 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 570992 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 94522 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 807817 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 95662 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 12168 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06973502 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06973479 | - | - |
| Whole Mouse Genome G4122A | A_51_P368427 | AK132706.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P368427 | AK016412.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115812 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06973502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934590 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08725800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934600 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08641904 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934721 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08725799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934631 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934624 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934616 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934696 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934735 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934685 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934689 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934622 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934730 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934683 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08487851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934733 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934605 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934628 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08636236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934686 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934692 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934592 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08799559 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934695 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934637 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08243002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934700 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08582885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08689497 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08800915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934584 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934671 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934810 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934726 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934585 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08717424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08633075 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934633 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934767 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08973967 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08897550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934765 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934699 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934607 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08552234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934653 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934701 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934702 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934687 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934797 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26934742 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 106350064 | - | - |

4930598N05Rik
Genomic Map