Assay Details
| Gene Symbol: Hivep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK040673.1 | - | 2 - 3 | 132 | 78 | |
| AK143444.1 | - | 2 - 3 | 127 | 78 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hivep2| Gene Symbol: Hivep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK040673.1 | - | 2 - 3 | 132 | 78 | |
| AK143444.1 | - | 2 - 3 | 127 | 78 | ||
Set Membership: |
|