Assay Details
| Gene Symbol: Myo1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178440.4 | NP_848534.2 | 5 - 6 | 737 | 86 | |
| GenBank mRNA | AK030825.1 | - | 5 - 6 | 738 | 86 | |
| AK036461.1 | - | 5 - 6 | 722 | 86 | ||
| AK042033.1 | - | 5 - 6 | 688 | 86 | ||
| AK088011.1 | - | 5 - 6 | 738 | 86 | ||
| AK146251.1 | - | 5 - 6 | 710 | 86 | ||
| AK152797.1 | - | 5 - 6 | 731 | 86 | ||
| AK159700.1 | - | 5 - 6 | 713 | 86 | ||
| AK171284.1 | - | 5 - 6 | 731 | 86 | ||
| AK171505.1 | - | 5 - 6 | 710 | 86 | ||
| BC113755.1 | - | 5 - 6 | 681 | 40061202 | 86 | |
| BC113954.1 | - | 5 - 6 | 681 | 40061200 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Myo1g
Genomic Map