Assay Details
| Gene Symbol: Atp2b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177236.4 | NP_796210.2 | 1 - 2 | 247 | 95 | |
| GenBank mRNA | AK045516.1 | - | 1 - 2 | 248 | 95 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_177236.4 transcripts.
Set Membership: |
|

Atp2b3
Genomic Map