Assay Details
| Gene Symbol: 1700046C09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045918.1 | 1 - 2 | 70 | 86 | ||
| GenBank mRNA | AK006701.1 | - | 1 - 2 | 48 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4930554G24 gene | 4930554G24Rik | -399.1kb | Sense | Chr. 11 - 14134143 - 14179087 on NCBI Build GRCm38 | |
| Downstream | POM121 membrane glycoprotein-like 12 | Pom121l12 | 1700054H16Rik | 26.0 | Anti-sense | Chr. 11 - 14599240 - 14600316 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

1700046C09Rik
Genomic Map