Assay Details
| Gene Symbol: 1700042O10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045178.1 | 3 - 4 | 398 | 70 | ||
| GenBank mRNA | AK006690.1 | - | 3 - 4 | 399 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | fidgetin-like 1 | Fignl1 | -74.0kb | Anti-sense | Chr. 11 - 11800288 - 11808995 on NCBI Build GRCm38 | |
| Downstream | growth factor receptor bound protein 10 | Grb10 |
5730571D09Rik, AI325020, Meg1, mKIAA0207 |
45.2kb | Anti-sense | Chr. 11 - 11930499 - 12037422 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1700042O10Rik
Genomic Map