Assay Details
| Gene Symbol: Gm11999 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK040327.1 | - | 1 - 2 | 114 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4930512M02 gene | 4930512M02Rik | -25.8kb | Sense | Chr. 11 - 11589223 - 11627818 on NCBI Build GRCm38 | |
| Downstream | fidgetin-like 1 | Fignl1 | 114.9kb | Sense | Chr. 11 - 11800288 - 11808995 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm11999
Genomic Map