Assay Details
| Gene Symbol: Ccdc157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164620.1 | NP_001158092.1 | 10 - 11 | 2118 | 103 | |
| NM_177616.3 | NP_808284.2 | 11 - 12 | 2254 | 103 | ||
| GenBank mRNA | AK076934.1 | - | 11 - 12 | 2182 | 103 | |
| AK133165.1 | - | 10 - 11 | 2119 | 103 | ||
| AK143216.1 | - | 9 - 10 | 1774 | 103 | ||
| AK220409.1 | - | 11 - 12 | 2235 | 103 | ||
| BC046392.1 | - | 4 - 5 | 997 | 103 | ||
| BC120737.1 | - | 8 - 9 | 1616 | 40129660 | 103 | |
| BC120739.1 | - | 8 - 9 | 1616 | 40129662 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc157
Genomic Map