Assay Details
| Gene Symbol: Pla2g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172791.2 | NP_766379.2 | 5 - 6 | 1309 | 55 | |
| GenBank mRNA | AK033572.1 | - | 5 - 6 | 1253 | 55 | |
| AK081075.1 | - | 5 - 6 | 1237 | 55 | ||
| BC079556.1 | - | 5 - 6 | 1309 | 30360278 | 55 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pla2g3
Genomic Map