Assay Details
| Gene Symbol: Htr1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285482.1 | NP_001272411.1 | 2 - 3 | 412 | 57 | |
| NM_001285483.1 | NP_001272412.1 | 2 - 3 | 234 | 57 | ||
| GenBank mRNA | AW487918.1 | - | 1 - 2 | 57 | ||
| BC103535.1 | - | 2 - 3 | 228 | 40039732 | 57 | |
| BC103536.1 | - | 2 - 3 | 410 | 40039733 | 57 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC103535.1, BC103536.1, NM_001285482.1, NM_001285483.1 transcripts.
Set Membership: |
|

Htr1d
Genomic Map