Assay Details
| Gene Symbol: Renbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164704.1 | NP_001158176.1 | 3 - 4 | 283 | 111 | |
| GenBank mRNA | AK167648.1 | - | 3 - 4 | 196 | 111 | |
| BC014821.1 | - | 3 - 4 | 219 | 4238244 | 111 | |
| BC035455.1 | - | 3 - 4 | 219 | 111 | ||
More Information
Set Membership: |
|

Renbp
Genomic Map