Assay Details
Gene Symbol: Fgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK036476.1 | - | 1 - 2 | 347 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK036476.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
