Assay Details
| Gene Symbol: Gopc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199272.1 | NP_001186201.1 | 2 - 3 | 681 | 90 | |
| GenBank mRNA | AK030427.1 | - | 2 - 3 | 277 | 90 | |
| AK030637.1 | - | 2 - 3 | 490 | 90 | ||
| AK032613.1 | - | 2 - 3 | 487 | 90 | ||
More Information
Set Membership: |
|

Gopc
Genomic Map