Assay Details
| Gene Symbol: 1700125G02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040651.1 | 2 - 3 | 426 | 69 | ||
| GenBank mRNA | AK007278.1 | - | 2 - 3 | 432 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Eph receptor A10 | Epha10 | A330090H18 | Anti-sense | Chr. 4 - 124881785 - 124917800 on NCBI Build GRCm38 | |
| Downstream | cell division cycle associated 8 | Cdca8 |
4831429J16Rik, AU044747, BOR, Borealin, D4Ertd421e, DasraB, MESRGP |
27.0kb | Sense | Chr. 4 - 124918465 - 124936917 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1700125G02Rik
Genomic Map