Assay Details
| Gene Symbol: Epm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010146.2 | NP_034276.2 | 2 - 3 | 491 | 98 | |
| GenBank mRNA | BY730431.1 | - | 2 - 3 | 492 | 98 | |
| CN532498.1 | - | 1 - 2 | 122 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epm2a
Genomic Map