Assay Details
| Gene Symbol: Mtrf1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175374.3 | NP_780583.1 | 4 - 5 | 844 | 94 | |
| GenBank mRNA | AK036424.1 | - | 4 - 5 | 748 | 94 | |
| AK078897.1 | - | 4 - 5 | 812 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mtrf1l
Genomic Map