Assay Details
| Gene Symbol: Nefh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010904.3 | NP_035034.2 | 1 - 2 | 1004 | 93 | |
| GenBank mRNA | AK122388.1 | - | 1 - 2 | 906 | 93 | |
| M35131.1 | - | 2 - 3 | 1027 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nefh
Genomic Map