Assay Details
| Gene Symbol: Emd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007927.3 | NP_031953.1 | 3 - 4 | 534 | 114 | |
| GenBank mRNA | AK146361.1 | - | 3 - 4 | 380 | 114 | |
| AK160531.1 | - | 3 - 4 | 379 | 114 | ||
| AK167852.1 | - | 3 - 4 | 377 | 114 | ||
| BC132131.1 | - | 3 - 4 | 346 | 40130408 | 114 | |
| BC132133.1 | - | 3 - 4 | 346 | 40130410 | 114 | |
| U73902.1 | - | 3 - 4 | 384 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Emd
Genomic Map