Assay Details
| Gene Symbol: Aldh8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178713.4 | NP_848828.1 | 2 - 3 | 334 | 81 | |
| GenBank mRNA | AF510322.1 | - | 2 - 3 | 343 | 81 | |
| AK050298.1 | - | 2 - 3 | 335 | 81 | ||
| AK143752.1 | - | 2 - 3 | 335 | 81 | ||
| BC038493.1 | - | 2 - 3 | 346 | 4223059 | 81 | |
More Information
Set Membership: |
|

Aldh8a1
Genomic Map