Assay Details
| Gene Symbol: Sgk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161845.2 | NP_001155317.2 | 2 - 3 | 729 | 71 | |
| GenBank mRNA | BC070401.1 | - | 2 - 3 | 729 | 5695396 | 71 |
More Information
Updated Mapping Information
Set Membership: |
|

Sgk1
Genomic Map