Assay Details
| Gene Symbol: Sgk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161845.2 | NP_001155317.2 | 5 - 6 | 963 | 65 | |
| NM_001161847.2 | NP_001155319.1 | 3 - 4 | 464 | 65 | ||
| NM_001161848.2 | NP_001155320.1 | 3 - 4 | 257 | 65 | ||
| NM_001161849.2 | NP_001155321.1 | 3 - 4 | 338 | 65 | ||
| NM_001161850.2 | NP_001155322.1 | 3 - 4 | 445 | 65 | ||
| NM_011361.3 | NP_035491.1 | 3 - 4 | 313 | 65 | ||
| GenBank mRNA | AF139638.1 | - | 3 - 4 | 297 | 65 | |
| AF205855.1 | - | 3 - 4 | 294 | 65 | ||
| AK086892.1 | - | 3 - 4 | 297 | 65 | ||
| AK132234.1 | - | 3 - 4 | 385 | 65 | ||
| AK146062.1 | - | 3 - 4 | 294 | 65 | ||
| AK159131.1 | - | 3 - 4 | 297 | 65 | ||
| AK167364.1 | - | 3 - 4 | 294 | 65 | ||
| BC005720.1 | - | 3 - 4 | 274 | 3594892 | 65 | |
| BC070401.1 | - | 5 - 6 | 963 | 5695396 | 65 | |
| BY736002.1 | - | 3 - 4 | 257 | 65 | ||
| CK626189.1 | - | 3 - 4 | 445 | 65 | ||
| CN709375.1 | - | 3 - 4 | 181 | 65 | ||
More Information
Set Membership: |
|

Sgk1
Genomic Map