Assay Details
| Gene Symbol: Tmem222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025667.3 | NP_079943.2 | 3 - 4 | 372 | 92 | |
| GenBank mRNA | AK017503.1 | - | 3 - 4 | 368 | 92 | |
| AK079189.1 | - | 3 - 4 | 360 | 92 | ||
| AK160188.1 | - | 3 - 4 | 373 | 92 | ||
| BC037609.1 | - | 3 - 4 | 333 | 5356647 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem222
Genomic Map