Assay Details
| Gene Symbol: Gpr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008154.3 | NP_032180.1 | 2 | 1687 | 120 | |
| GenBank mRNA | AK140302.1 | - | 2 | 1658 | 120 | |
| AK140464.1 | - | 2 | 1641 | 120 | ||
| AK158917.1 | - | 2 | 1650 | 120 | ||
| D21062.1 | - | 1 | 2062 | 120 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gpr3
Genomic Map