Assay Details
| Gene Symbol: Sytl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031393.2 | NP_113570.2 | 1 - 2 | 102 | 113 | |
| GenBank mRNA | AA856092.1 | - | 1 - 2 | 102 | 111 | |
| AB050741.1 | - | 1 - 2 | 81 | 113 | ||
| AK078868.1 | - | 1 - 2 | 90 | 113 | ||
| AK131162.1 | - | 1 - 2 | 59 | 113 | ||
| AK154176.1 | - | 1 - 2 | 97 | 113 | ||
| BC005623.1 | - | 1 - 2 | 102 | 3155535 | 113 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB050741.1, BC005623.1, NM_031393.2 transcripts.
Set Membership: |
|

Sytl1
Genomic Map