Assay Details
| Gene Symbol: Sytl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB050741.1 | - | 5 - 6 | 582 | 63 | |
| AK154176.1 | - | 5 - 6 | 598 | 63 | ||
| BC005623.1 | - | 5 - 6 | 603 | 3155535 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sytl1
Genomic Map