Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400030478.mm.1 |
TC0400003717.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400030478.mm.1 |
NM_001285505.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030478.mm.1 |
NM_001285506.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030478.mm.1 |
NM_009097.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030480.mm.1 |
TC0400003717.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400030480.mm.1 |
NM_001285506.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030480.mm.1 |
NM_009097.5 |
Refseq |